Sodium diethyldithiocarbamate trihydrate, J.T.Baker®

Warning

Synonyms: Diethyldithiocarbamic acid sodium salt trihydrateN,N-Diethyldithiocarbamic acid sodium salt trihydrate(Diethyldithiocarbamato-S,S')sodium trihydrate

BAKR8624-01EA 207 SGD
BAKR8624-01
Sodium diethyldithiocarbamate trihydrate, J.T.Baker®
Sodium diethyldithiocarbamate trihydrate

Lot analysis on label.

Formula: (C₂H₅)₂NCSSNa·3H₂O
MW: 225,31 g/mol
Melting Pt: 95…98,5 °C
Density: 1,1 g/cm³ (20 °C)
MDL Number: MFCD00150617
CAS Number: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9,III
Merck Index: 12,03443

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Solubility in H₂O Passes Test
Sodium (as Na₂SO₄) 30.5 - 32.5 %
Sensitivity for Copper Passes Test
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR