Laboratory tables, Contour™
VWRUCFTM3630R30DEA 1350 SGD
VWRUCFTM3630R30D VWRUVFT-3630-DRT
Laboratory tables, Contour™
Made with welded leg assemblies connected to steel apron rails with metal fasteners, Contour laboratory tables provide a stable work surface.
  • Durable, economical resin worktop is smooth and self-extinguishing
  • Heat-resistant and contains no asbestos
  • Low-glare surface that is easy to clean and maintain
  • Lockable casters are available
The rails for the laboratory tables are 102 mm high and formed of 16-gauge steel. Table legs are made of 51 mm square welded tubing to provide a stable, solid workplace. Leg assemblies include levelling bolts, stretchers, and stringers for better support.
Colour: Light neutral
Ordering information: Table frames and table tops should be ordered separately.