Compact muffle furnaces, LE series
Supplier: Nabertherm
NABELE021K17NEA 2680 SGD
NABELE021K17N NABELE141K17N NABELE061K17N NABELE012K17N
Compact muffle furnaces, LE series
With an excellent price/performance ratio, these compact, lightweight muffle furnaces are ideal for many laboratory applications. Units feature double walled, rust-free, textured stainless steel exteriors which ensure low external temperatures and high stability. The heating elements are encased in quartz glass tubes making them reliable for a variety of applications. The furnaces have Tmax 1100 °C, working temperature of 1050 °C. Models have an exhaust air outlet in the rear wall of the furnace and a silent solid state power control relay.
  • Heating on two sides by means of heating elements in quartz glass tubes
  • Maintenance friendly replacement of heating elements and insulation
  • Dual shell housing for low external temperatures and high stability
  • Controller is mounted to the side of the door for larger models but under the door for the LE 1/11, LE 2/11 and LE 4/11
  • Fold down door can also be used as a work platform
  • Non-classified insulation avoids the potential risk of cancer
Delivery information: All models supplied with digital R 7 controller with one holding temperature. All units are supplied with plugs and these models require a 230 V single phase power supply.
Caution: Maximum continuous operating temperature should be 50 °C below the max. temp. of the model shown in the table above.