Dichromate, ion chromatography standard solution, Specpure®, Cr₂O₇-² 1000 µg/ml

Supplier: Thermo Fisher Scientific
ALFA42565.APEA 263 SGD
ALFA42565.AP ALFA42565.AE
Dichromate, ion chromatography standard solution, Specpure®, Cr₂O₇-² 1000 µg/ml
Molecular Formula: Na₂Cr₂O₇ in H₂O
Physical Form: Liquid
MDL No.: MFCD01093393
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR