Polyethylene, chlorinated
Supplier: Thermo Scientific
ALFA44146.30EA
0
SGD
ALFA44146.30
ALFA44146.A1
Polyethylene, chlorinated
Polyethylene
Formula:
(CH₂CH₂)n Storage Temperature: Ambient |
MDL Number:
MFCD00084424 CAS Number: 64754-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...