Ethanolamine benzoate hydrochloride ≥98%
Supplier: Thermo Scientific
|
Warning
|
ALFAH25864.14EA
281
SGD
ALFAH25864.14
Ethanolamine benzoate hydrochloride ≥98%
Ethanolamine benzoate hydrochloride
Formula:
C₉H₁₁NO₂·HCl MW: 201,65 g/mol Melting Pt: 141…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD09953460 CAS Number: 545375-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...