Chlorobis[4-(trifluoromethyl)phenyl]phosphine 97%

Supplier: Thermo Scientific
Danger

ALFAH25888.06EA 651 SGD
ALFAH25888.06 ALFAH25888.03
Chlorobis[4-(trifluoromethyl)phenyl]phosphine 97%
Chlorobis[4-(trifluoromethyl)phenyl]phosphine
Formula: C₁₄H₈ClF₆P
MW: 356,64 g/mol
Boiling Pt: 110 °C (0,4 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD04972304
CAS Number: 13685-24-0
EINECS: 000-000-0
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR