Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Bromononanophenone ≥98%

Supplier: Alfa Aesar

Purity: 98%
CAS No.: 126334-26-7
Molecular Formula: C15H21BrO
Formula Weight: 297.24
Melting Point: 65-69°
Boiling Point: 138-140°/0.1mm
MDL No.: MFCD09953463

Formula: C₁₅H₂₁BrO
MDL Number: MFCD09953463
CAS Number: 126334-26-7

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Pk VWR Catalog Number Quantity
25 g ALFAH25900.14
Supplier Article Number:: H25900.14
5 g ALFAH25900.06
Supplier Article Number:: H25900.06
Expand All / Collapse All


Melting point 65-69°C
Formula weight 297.24
CAS No. 126334-26-7
Purity 98%
Boiling point 138-140°/0.1mm
Molecular formula C15H21BrO
MDL No. MFCD09953463
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty