(E)-3,4-Dihydroxybenzylideneacetone 97%

Supplier: Thermo Scientific
Warning

ALFAH25901.06EA 783 SGD
ALFAH25901.06
(E)-3,4-Dihydroxybenzylideneacetone 97%
(E)-3,4-Dihydroxybenzylideneacetone
Formula: C₁₀H₁₀O₃
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00916691
CAS Number: 123694-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR