Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-1-Boc-4,4-diethoxypiperidine-2-carboxylic acid ≥98%

Supplier: Alfa Aesar

Purity: 98%
CAS No.: 661458-35-1
Molecular Formula: C15H27NO6
Formula Weight: 317.38
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4620
MDL No.: MFCD08056311


Formula: C₁₅H₂₇NO₆
MDL Number: MFCD08056311
CAS Number: 661458-35-1


 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
100 mg ALFAH25905.MC
Supplier Article Number:: H25905.MC
500 mg ALFAH25905.ME
Supplier Article Number:: H25905.ME
Expand All / Collapse All

SPECIFICATIONS

Formula weight 317.38
CAS No. 661458-35-1
Purity 98%
Refractive index 1.4620
Storage temperature 2°C to 8°C
Molecular formula C15H27NO6
MDL No. MFCD08056311
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty