Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(-)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate

Supplier: Alfa Aesar

Molecular Formula: C15H30N2O4.C4H4O4
Formula Weight: 418.49
MDL No.: MFCD09953464



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
100 mg ALFAH25907.MC
Supplier Article Number:: H25907.MC
500 mg ALFAH25907.ME
Supplier Article Number:: H25907.ME
Expand All / Collapse All

SPECIFICATIONS

Formula Weight 418.49
Molecular Formula C15H30N2O4.C4H4O4
MDL No. MFCD09953464
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty