Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Albendazole sulfone

Supplier: Alfa Aesar

CAS No.: 75184-71-3
Molecular Formula: C12H15N3O4S
Formula Weight: 297.33
Melting Point: 290-292°
MDL No.: MFCD00600775


Formula: C₁₂H₁₅N₃O₄S
MDL Number: MFCD00600775
CAS Number: 75184-71-3

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g ALFAH25917.03
Supplier Article Number:: H25917.03
Expand All / Collapse All

SPECIFICATIONS

Melting point 290-292°C
Formula weight 297.33
CAS No. 75184-71-3
Molecular formula C12H15N3O4S
MDL No. MFCD00600775
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty