6-Methylbenzimidazole-5-carboxylic acid hydrochloride 97%
Supplier: Thermo Scientific
|
Warning
|
ALFAH25926.09EA
494
SGD
ALFAH25926.09
6-Methylbenzimidazole-5-carboxylic acid hydrochloride 97%
6-Methylbenzimidazole-5-carboxylic acid hydrochloride
Formula:
C₉H₈N₂O₂·HCl MW: 212,64 g/mol Melting Pt: 315 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD09909565 CAS Number: 10351-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...