Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7-triacetate
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic Acid Tri-tert-butyl Ester
, Tri-tert-butyl 2,2',2''-(1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate
ALFAH26736.MDEA
509
SGD
ALFAH26736.MD
Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7-triacetate
Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7-triacetate
Formula:
C₂₆H₅₀N₄O₆ MW: 514,71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05865261 CAS Number: 122555-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...