Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4,8-Tri-Boc-1,4,8,11-tetraazacyclotetradecane

Supplier: Alfa Aesar

CAS No.: 170161-27-0
Molecular Formula: C25H48N4O6
Formula Weight: 500.68
Melting Point: 48-54°
MDL No.: MFCD09263315


Formula: C₂₅H₄₈N₄O₆
MDL Number: MFCD09263315
CAS Number: 170161-27-0


 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
250 mg ALFAH26741.MD
Supplier Article Number:: H26741.MD
Expand All / Collapse All

SPECIFICATIONS

Melting point 48-54°C
Formula weight 500.68
CAS No. 170161-27-0
Molecular formula C25H48N4O6
MDL No. MFCD09263315
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty