Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Methoxyphenyl)thioacetamide ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 60759-02-6
Molecular Formula: C9H11NOS
Formula Weight: 181.27
Melting Point: 139-142°
MDL No.: MFCD00278203

 
  Formula: C₉H₁₁NOS
MDL Number: MFCD00278203
CAS Number: 60759-02-6
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g ALFAH26742.03
5 g ALFAH26742.06
Expand All / Collapse All

SPECIFICATIONS

Melting Point 139-142°C
Formula Weight 181.27
CAS No. 60759-02-6
Purity 97%
Molecular Formula C9H11NOS
MDL No. MFCD00278203
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty