Pyrazine-2-carbothioamide 97%
Supplier: Thermo Scientific
|
Warning
|
ALFAH26750.03EA
231
SGD
ALFAH26750.03
ALFAH26750.06
ALFAH26750.MD
Pyrazine-2-carbothioamide 97%
Pyrazine-2-carbothioamide
Formula:
C₅H₅N₃S MW: 139,18 g/mol Melting Pt: 198…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173663 CAS Number: 4604-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...