Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraethyl 1,4,8,11-Tetraazacyclotetradecane-1,4,8,11-tetraacetate

Supplier: Alfa Aesar
Synonyms: 1,4,8,11-Tetraazacyclotetradecane-1,4,8,11-tetraacetic Acid Tetraethyl Ester

CAS No.: 126320-57-8
Molecular Formula: C26H48N4O8
Formula Weight: 544.69
Melting Point: 87-89°
MDL No.: MFCD09263316

Formula: C₂₆H₄₈N₄O₈
MW: 544,69 g/mol
CAS Number: 126320-57-8

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g ALFAH26755.03
Supplier Article Number:: H26755.03
250 mg ALFAH26755.MD
Supplier Article Number:: H26755.MD
Expand All / Collapse All

SPECIFICATIONS

Melting Point 87-89°C
Formula Weight 544.69
CAS No. 126320-57-8
Molecular Formula C26H48N4O8
MDL No. MFCD09263316
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty