Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Chlorophenylsulphonyl)thioacetamide ≥97%

Supplier: Alfa Aesar
Danger



Formula: C₈H₈ClNO₂S₂
MW: 249,73
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00052824
CAS Number: 59865-87-1
EINECS: None

 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g ALFAH26806.03
Supplier Article Number:: H26806.03
250 mg ALFAH26806.MD
Supplier Article Number:: H26806.MD
5 g ALFAH26806.06
Supplier Article Number:: H26806.06
Expand All / Collapse All

SPECIFICATIONS

Melting point 186-190°C
Formula weight 249.73
CAS No. 59865-87-1
Purity 97%
Molecular formula C8H8ClNO2S2
MDL No. MFCD00052824
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty