Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Biphenyl-4-yl-piperazine ≥95%

Supplier: Alfa Aesar

Purity: 95%
CAS No.: 105242-10-2
Molecular Formula: C16H18N2
Formula Weight: 238.34
Melting Point: 156-159°
MDL No.: MFCD05864686


Formula: C₁₆H₁₈N₂
CAS Number: 105242-10-2

 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g ALFAH26820.03
Supplier Article Number:: H26820.03
250 mg ALFAH26820.MD
Supplier Article Number:: H26820.MD
Expand All / Collapse All

SPECIFICATIONS

Melting point 156-159°C
Formula weight 238.34
CAS No. 105242-10-2
Purity 95%
Molecular formula C16H18N2
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty