Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-sec-Butyl-4,5-dimethyl-3-thiazoline ≥98%

Supplier: Alfa Aesar

Purity: 98%
CAS No.: 65894-82-8
Molecular Formula: C9H17NS
Formula Weight: 171.30
Flash Point: >70°(158°F)
Refractive Index: 1.4920
Boiling Point: 71.00°/4.00mm
MDL No.: MFCD01680858

 
  Formula: C₉H₁₇NS
MDL Number: MFCD01680858
CAS Number: 65894-82-8
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
10 g ALFAH26870.09
1 g ALFAH26870.03
Expand All / Collapse All

SPECIFICATIONS

Formula Weight 171.30
CAS No. 65894-82-8
Purity 98%
Refractive Index 1.4920
Flash Point >70°C(158°F)
Boiling Point 71.00°/4.00mm
Molecular Formula C9H17NS
MDL No. MFCD01680858
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty