3-(3-Benzyloxy-4-methoxyphenyl)propionic acid 96%

Supplier: Thermo Scientific
Warning

ALFAH33108.03EA 0 SGD
ALFAH33108.03
3-(3-Benzyloxy-4-methoxyphenyl)propionic acid 96%
3-(3-Benzyloxy-4-methoxyphenyl)propionic acid
Formula: C₁₇H₁₈O₄
MW: 286,33 g/mol
Melting Pt: 121…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00227803
CAS Number: 36418-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR