4,6-Dihydroxy-2-(trifluoromethyl)pyrimidine 97%

Warning

Synonyms: 2-Trifluoromethyl-pyrimidine-4,6-diol

ALFAH33364.03EA 221 SGD
ALFAH33364.03
4,6-Dihydroxy-2-(trifluoromethyl)pyrimidine 97%
4,6-Dihydroxy-2-(trifluoromethyl)pyrimidine
Formula: C₅H₃F₃N₂O₂
MW: 180,09 g/mol
Melting Pt: 254…256 °C
Storage Temperature: Ambient
MDL Number: MFCD02678234
CAS Number: 672-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR