Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4,4,4,4',4',4'-Hexafluorovaline, ethyl ester
ALFAH33837.03EA
0
SGD
ALFAH33837.03
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate 95%
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate
Formula:
C₇H₉F₆NO₂ MW: 253,15 g/mol Boiling Pt: 70…72 °C (10 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00040845 CAS Number: 78185-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...