3-Nitroisonicotinic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Nitropyridine-4-carboxylic acid

ALFAH34293.MDEA 0 SGD
ALFAH34293.MD ALFAH34293.03
3-Nitroisonicotinic acid 97%
3-Nitroisonicotinic acid
Formula: C6H4N2O4
MW: 168,11 g/mol
Melting Pt: ∼ 220 °CC (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04114244
CAS Number: 59290-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR