(R)-(-)-4-(3-Indolylmethyl)-2-oxazolidinone 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(R)-4-((1H-Indol-3-yl)methyl)oxazolidin-2-one
ALFAH51075.03EA
370
SGD
ALFAH51075.03
(R)-(-)-4-(3-Indolylmethyl)-2-oxazolidinone 98%
(R)-(-)-4-(3-Indolylmethyl)-2-oxazolidinone
Formula:
C₁₂H₁₂N₂O₂ MW: 216,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01863583 CAS Number: 157636-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...