Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

S-Methyl pyridine-4-carbothioimidate hydriodide ≥96%

Supplier: Alfa Aesar
Warning



Formula: C₇H₈N₂S·HI
MW: 280,13
Melting Pt: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD12026517
CAS Number: 1185165-63-2

 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g ALFAH51774.03
Supplier Article Number:: H51774.03
5 g ALFAH51774.06
Supplier Article Number:: H51774.06
Expand All / Collapse All

SPECIFICATIONS

Melting point 182-186°C
Formula weight 280.13
Purity 96%
Molecular formula C7H8N2S.HI
MDL No. MFCD12026517
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty