Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzene-1,3-dithiocarboxamide ≥97%

Supplier: Alfa Aesar
Danger



Formula: C₈H₈N₂S₂
MW: 196,28
Melting Pt: 198…200 °C
Storage Temperature: Ambient
MDL Number: MFCD16293899
CAS Number: 3030-54-4
ADR: 6.1,III

 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g ALFAH51853.03
Supplier Article Number:: H51853.03
5 g ALFAH51853.06
Supplier Article Number:: H51853.06
Expand All / Collapse All

SPECIFICATIONS

Formula weight 196.28
Purity 97%
Molecular formula C8H8N2S2
MDL No. MFCD16293899
UN No. 2811
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty