4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester 95%
Supplier: Thermo Scientific
Synonyms:
Methyl 4-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethynyl)benzoate
ALFAH51900.03EA
152
SGD
ALFAH51900.03
4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester 95%
4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester
Formula:
C₂₂H₂₃BO₄ MW: 362,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16294527 CAS Number: 1448637-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...