Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl 4'-chloro[1,1'-biphenyl]-4-carboxylate ≥95%

Supplier: Alfa Aesar

Purity: 95%
CAS No.: 89901-02-0
Molecular Formula: C14H11ClO2
Formula Weight: 246.69
MDL No.: MFCD04117794


Formula: C₁₄H₁₁ClO₂
MW: 246,7 g/mol
MDL Number: MFCD04117794
CAS Number: 89901-02-0

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g ALFAH51910.03
Supplier Article Number:: H51910.03
5 g ALFAH51910.06
Supplier Article Number:: H51910.06
Expand All / Collapse All

SPECIFICATIONS

Formula weight 246.69
CAS No. 89901-02-0
Purity 95%
Molecular formula C14H11ClO2
MDL No. MFCD04117794
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty