Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(4-Methoxyphenylethynyl)benzeneboronic acid pinacol ester ≥95%

Supplier: Alfa Aesar



Formula: C₂₁H₂₃BO₃
MW: 334,22
Storage Temperature: Ambient
MDL Number: MFCD16294533
CAS Number: 1190376-24-9

 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g ALFAH51914.03
Supplier Article Number:: H51914.03
5 g ALFAH51914.06
Supplier Article Number:: H51914.06
Expand All / Collapse All

SPECIFICATIONS

Formula weight 334.22
CAS No. 1190376-24-9
Purity 95%
Molecular formula C21H23BO3
MDL No. MFCD16294533
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty