LDS-sample buffer (4X), reducing

Supplier: Thermo Fisher Scientific
ALFAJ61942.ADEA 0 SGD
ALFAJ61942.AD ALFAJ61942.AC
LDS-sample buffer (4X), reducing
Physical Form: Viscous liquid
Appearance: Blue
UN No.:UN3082
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR