Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gelatin blocking buffer, 1% in PBS

Supplier: Alfa Aesar
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD01634945



 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 l ALFAJ62755.K2
Supplier Article Number:: J62755.K2
500 ml ALFAJ62755.AP
Supplier Article Number:: J62755.AP

SPECIFICATIONS

Storage temperature -30°C to -10°C
Physical form Liquid
Appearance Colourless
MDL No. MFCD01634945
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty