Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Casein blocking buffer, 3% in PBS, with 0.02% sodium azide

Supplier: Alfa Aesar
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD01634587
Applications: For Western Blots and ELISA



 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
250 ml ALFAJ63453.AK
Supplier Article Number:: J63453.AK
500 ml ALFAJ63453.AP
Supplier Article Number:: J63453.AP

SPECIFICATIONS

Storage temperature -30°C to -10°C
Physical form Liquid
MDL No. MFCD01634587
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty