Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Lithium Standards

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"11"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Lithium 10 mg/l ARISTAR mono-element Standard for ICP-MS 1 * 100 ml
Supplier:  VWR Chemicals
Description:   Li in H₂O tr.HNO₃
UOM:  1 * 100 ml
Supplier:  VWR Chemicals
Description:   Lithium standard solution, 1,000 mg/l Li in dil. nitric acid AVS TITRINORM® standard for AAS
Supplier:  VWR Chemicals
Description:   Li₂CO₃ in HNO₃ 2 - 5%
Supplier:  VWR Chemicals
Description:   Li in H₂O tr.HNO₃
UOM:  1 * 100 ml
Supplier:  VWR Chemicals
Description:   Li₂CO₃ in HNO₃ 2 - 5%
UOM:  1 * 500 ml
Supplier:  VWR Chemicals
Description:   Lithium 1000 mg/l in water standard for ion chromatography
UOM:  1 * 100 ml
Supplier:  Avantor Performance Materials
Supplier Article Number:: 6456-04
Description:   Lithium standard solution for AAS.
UOM:  1 * 150 ml
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR at sales@sg.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
1 - 8  of 11
  1  2  Next