Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Molybdenum Standards

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Molybdenum standard solution, 1,000 mg/l Mo in water AVS TITRINORM® standard for AAS
UOM:  1 * 500 ml
Supplier:  VWR Chemicals
Description:   (NH₄)₂MoO₄ in HNO₃ tr. HF
Supplier:  VWR Chemicals
Description:   Molybdenum 1000 ppm ARISTAR mono-element Standard for ICP-MS 1 * 100 ml
UOM:  1 * 100 ml
Supplier:  VWR Chemicals
Description:   (NH₄)₂MoO₄ in HNO₃ tr. HF
UOM:  1 * 100 ml
Supplier:  Alfa Aesar
Description:   Molybdenum standard solution, 10,000 mg/l Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo) Specpure® standard for ICP
Supplier:  Alfa Aesar
Description:   Molybdenum standard solution, 1,000 mg/l Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo) Specpure® standard for ICP
Supplier:  Alfa Aesar
Description:   Molybdenum standard solution, 1,000 mg/l Mo in ammonium hydroxide solution (max. 1%) (from (NH4)2MoO4) Specpure® standard for ICP
Supplier:  Alfa Aesar
Description:   Molybdenum standard solution, 1,000 mg/l Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo) Specpure® standard for AAS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR at sales@sg.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
1 - 8  of 9
  1  2  Next