Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Rhenium Standards

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"7"
  List View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Re in H₂O tr. HNO₃
UOM:  1 * 100 ml
Supplier:  VWR Chemicals
Description:   Re in H₂O tr. HNO₃
Supplier:  Alfa Aesar
Description:   Rhenium standard solution, 1,000 mg/l Re in 5% nitric acid (from Re) Specpure® standard for ICP
Supplier:  Alfa Aesar
Description:   Rhenium standard solution, 1,000 mg/l Re in 5% nitric acid (from Re) Specpure® standard for AAS
Supplier:  Alfa Aesar
Description:   Rhenium standard solution, 10,000 mg/l Re in 5% nitric acid (from Re) Specpure® standard for ICP
Supplier:  Panreac Química SLU
Supplier Article Number:: 775991.1208
Description:   Rhenium standard solution, 10,000 mg/l Re in water (from (NH4)ReO4) standard for ICP
UOM:  1 * 100 ml
Supplier:  Panreac Química SLU
Supplier Article Number:: 765925.1208
Description:   Rhenium standard solution, 1,000 mg/l Re in water (from (NH4)ReO4) standard for ICP
UOM:  1 * 100 ml
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR at sales@sg.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
1 - 7  of 7
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping