Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

You Searched For:

Thallium Standards

10  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
 
 
SearchResultCount:"10"
  List View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Thallium 10 mg/l ARISTAR mono-element Standard for ICP-MS 1 * 100 ml
Supplier:  VWR Chemicals
Description:   Tl in HNO₃ 2 - 5%
UOM:  1 * 100 ml
Supplier:  VWR Chemicals
Description:   Tl in HNO₃ 2 - 5%
UOM:  1 * 100 ml
Supplier:  Alfa Aesar
Description:   Thallium standard solution, 1,000 mg/l Tl in 5% nitric acid (from Tl) Specpure® standard for AAS
Supplier:  Alfa Aesar
Description:   Thallium standard solution, 10,000 mg/l Tl in 5% nitric acid (from Tl) Specpure® standard for ICP
Supplier:  Alfa Aesar
Description:   Thallium standard solution, 1,000 mg/l Tl in 5% nitric acid (from Tl) Specpure® standard for ICP
Supplier:  Alfa Aesar
Supplier Article Number:: 45306.18
Description:   Thallium standard dissolved in oil, 1,000 µg/g Tl in mineral oil Specpure® for wear metal analysis
UOM:  1 * 50 g
Supplier:  Alfa Aesar
Supplier Article Number:: 45288.AD
Description:   Thallium 203, ²⁰ Tl, plasma standard solution, Specpure®, ²⁰ Tl 10µg/ml 1 * 50 ml
UOM:  1 * 50 ml
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR at sales@sg.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
1 - 8  of 10
  1  2  Next