Would you like to log in again?
Bomb for gum stability, PYREX®
Supplier: Corning Life Sciences
PYREX®
Bomb with cover made from Pyrex® borosilicate glass to determine gum stability in gasoline.
  • Wide mouth with pourout style top
  • Cover to prevent condensed vapours from contaminating the sample
  • No interference with the free flow of oxygen
Certifications: ASTM D-525
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Gum bomb (O.Ø×H = 50×105 mm) 1 CORN7732
Supplier Article Number:: 7732
ACCESSORIES
Expand All / Collapse All
Accessories
Description Pk VWR Catalog Number Quantity
Cover for gum bomb 1 CORN7732-CO
Supplier Article Number:: 7732-CO
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty