Disposable particle filtering respirators, FFP2, uvex silv-air 7200
Supplier: Uvex
UVEX8707.220EA 131 SGD
UVEX8707.200 UVEX8707.210 UVEX8707.220
Disposable particle filtering respirators, FFP2, uvex silv-air 7200
Class FFP2 masks are particularly suitable for use in mining as well as in the metalworking industry, in shipyards and in wood processing. All FFP2 respirators provide reliable protection against dust of up to ten times the limit value (TLV) and are available with or without valves.
  • New optimised shape
  • Soft fabric edges for improved comfort
  • Seamless, continuous headband to ensure mask is comfortable and secure
  • Exhalation valve reduces heat and moisture under the mask
  • Mask fulfils the dolomite dust test requirements
  • Optimal nose section design of the moulded masks ensures compatibility with safety goggles (moulding on the left and right-hand side of the nose)
  • Carbon version filters unpleasant odours (gases and vapours below the threshold value)
Class FFP2 masks are particularly suitable for use in mining as well as in the metalworking industry, in shipyards and in wood processing. All FFP2 respirators provide reliable protection against dust of up to ten times the limit value (TLV) and are available with or without valves.
  • New optimised shape
  • Soft fabric edges for improved comfort
  • Seamless, continuous headband to ensure mask is comfortable and secure
  • Exhalation valve reduces heat and moisture under the mask
  • Mask fulfils the dolomite dust test requirements
  • Optimal nose section design of the moulded masks ensures compatibility with safety goggles (moulding on the left and right-hand side of the nose)
  • Carbon version filters unpleasant odours (gases and vapours below the threshold value)
Caution: *One size fits all
*One size fits all