Statistical printer for VWR balances
VWRI611-3623EA 1030 SGD
VWRI611-3623
Statistical printer for VWR balances
The statistical printer is a bidirectional thermal printer that can be used with laboratory balances. It can print weight values and, when requested, a full statistical analysis of the values including: Number of values, mean value, sum, minimum, maximum, standard deviation and more. The printer can also record and print date/time and calibration details in compliance with GLP.
  • Count and total of all measurements, minimum, maximum values, range (max-min)
  • Average value, standard deviation
  • GLP record keeping of balance calibration with date/time print
  • Available units: mg, g, kg, pcs
  • Languages: English, French, German
Delivery information: Supplied with connection cable for direct connection to a balance.
Caution: Printer can not be used with moisture analysers or balances with a graphic display/touchscreen.