Copper single element plasma standard solutions,1000 µg/ml Cu, BAKER INSTRA-ANALYZED® J.T. Baker®
BAKR5713-04EA 248 SGD
BAKR5713-04
Copper single element plasma standard solutions,1000 µg/ml Cu, BAKER INSTRA-ANALYZED® J.T. Baker®
Single element plasma standards are used for calibrating elements and also to trace impurities.
Certifications: NIST traceable.