This non-DEHP tubings are ideal for most inorganic chemicals in the laboratory. It is non-oxidising and non-contaminating, and less permeable than rubber tubing. The glassy, smooth inner bore helps prevent build up to facilitate cleaning, and coils are marked at 30,4 cm intervals for easy measuring.
- Outstanding chemical resistance
- Slips easily over fittings and grips securely for simple lab set-ups
- Contains no BPA or phthalates
- Lot-to-lot consistency for reproducible results
- Ideal for condensers, incubators, desiccators, gas lines, and drain lines
Concentrated acids and concentrated bases tend to react with the bio-based Tygon® E-3603 more than with the Tygon® R-3603. For these conditions, the Tygon® LMT-55 can be used as an alternative product.
Temperature resistance: −46 to +74 °C
Hardness (Shore durometer A): 56
Density: 1,18
Certifications: Complies with FDA, NSF 51 & 61, meets USP Class VI