VWR® UV-3000PC, Uv/vis spectrophotometer
Supplier: VWR
VWR®
VWRU10037-438EA 8000 SGD
VWRU10037-438
VWR® UV-3000PC, Uv/vis spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
This spectrophotometer is an advanced single beam designed PC model that differs in bandwidth and wavelength accuracy by providing efficient performance for measurements.
  • Fixed or variable bandwidths
  • Sealed and solvent-resistant tactile keypad with alpha-numeric entry for user file names and user entry of units
  • Full A4 print-out of graphs and tables of results
  • Real-time clock for date and time stamping of results
  • Data can be saved by USB memory device directly
Suitable for clinical, pharmaceutical and biochemical laboratory applications, as well as routine applications including quantitative analysis, kinetics, wavelength scan, multi-wavelength and DNA/protein analysis.
Delivery information: Supplied with four optical glass cells, two quartz cells, dust cover, USB cable, 8 Gb USB stick, UV-Vis analyst software for PC control and four-position cell holder (10 mm ).