You Searched For: Ninhydrin


11  results were found

SearchResultCount:"11"

Sort Results

List View Easy View (new)

Rate These Search Results

Catalog Number: (VWRC87879.180)
Supplier: VWR Chemicals
Description: A 4 g/l solution in a mixture of 5 volumes of anhydrous acetic acid R and 95 volumes of butanol R.
UOM: 1 * 100 mL

Catalog Number: (VWRC87877.180)
Supplier: VWR Chemicals
Description: Ninhydrin 1,7% in ethanol 83.3% (v/v) Reag. Ph. Eur. 1058304
UOM: 1 * 100 mL

Supplier: VWR Chemicals
Description: Ninhydrin, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Catalog Number: (VWRC30960.226)
Supplier: VWR Chemicals
Description: Ninhydrin 0,5% in 1-butanol, spray reagent for TLC
UOM: 1 * 1 Each

Catalog Number: (VWRC87713.180)
Supplier: VWR Chemicals
Description: Ninhydrin 0.2% in 1-butanol : acetic acid (95:5 v:v)
UOM: 1 * 100 mL

Catalog Number: (VWRC87878.180)
Supplier: VWR Chemicals
Description: Ninhydrin R (3 g) in 100 ml of a 45,5 g/l solution of sodium metabisulphite R.
UOM: 1 * 100 mL

Supplier: Thermo Scientific
Description: Ninhydrin ACS
Supplier: Thermo Scientific
Description: Ninhydrin ≥99%
Catalog Number: (ICNA0210245090)
Supplier: MP Biomedicals
Supplier Article Number:: 0210245090
Description: Ninhydrin
UOM: 1 * 500 g


Catalog Number: (APPCA0902.0100)
Supplier: PanReac AppliChem
Supplier Article Number:: A0902.0100
Description: Ninhydrin
UOM: 1 * 100 g


Catalog Number: (BEAN219340-25G)
Supplier: BEANTOWN CHEMICAL
Supplier Article Number:: 219340-25G
Description: Ninhydrin ACS
UOM: 1 * 25 g


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR at sales@sg.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
1 - 11 of 11
no targeter for Bottom