You Searched For: Potassium dihydrogen phosphate


36  results were found

SearchResultCount:"36"

Sort Results

List View Easy View (new)

Rate These Search Results

Supplier: Avantor
Description: Potassium dihydrogen phosphate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: MACRON (A AVANTOR BRAND)
Description: Potassium dihydrogen phosphate 98.0-100.5% (dried basis, RT), crystals, GenAR® NF, Macron Fine Chemicals™
Supplier: VWR Chemicals
Description: Potassium dihydrogen phosphate, anhydrous ≥99% ACS
Supplier: Avantor
Description: Maximise the stability of your pharmaceutical formulations and get predictable pH control with reliable, proven buffering agents from Avantorâ„¢ Perform...
Supplier: VWR Chemicals
Description: Potassium dihydrogen phosphate Ph. Eur., NF, BP endotoxin tested
Supplier: VWR Chemicals
Description: Potassium dihydrogen phosphate 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: Avantor
Description: Potassium dihydrogen phosphate, ULTREX®, Ultrapure, J.T.Baker®
Catalog Number: (BAKR3248-X2)
Supplier: Avantor
Description: Maximise the stability of your pharmaceutical formulations and get predictable pH control with reliable, proven buffering agents from Avantorâ„¢ Perform...
UOM: 1 * 1 KIT

Catalog Number: (VWRC87901.290)
Supplier: VWR Chemicals
Description: Potassium dihydrogen phosphate 0.2 mol/l in aqueous solution Reag. Ph. Eur. 1069601
UOM: 1 * 1 L

Supplier: VWR Chemicals
Description: Potassium dihydrogen phosphate 98.0-100.5%, crystallised Ph. Eur.
Catalog Number: (BAKR1737.1000)
Supplier: Avantor
Description: Potassium dihydrogen phosphate, BAKER, Laboratory Reagent, J.T.Baker®
UOM: 1 * 1 kg

MSDS


Supplier: MACRON (A AVANTOR BRAND)
Description: Potassium dihydrogen phosphate ≥99.0% (by acidimetry), crystals, AR® ACS, Macron Fine Chemicals™
Supplier: Avantor
Description: Crystals. For LC and molecular biology. Lot analysis on label.
Supplier: Avantor
Description: Maximise the stability of your pharmaceutical formulations and get predictable pH control with reliable, proven buffering agents from Avantorâ„¢ Perform...
Supplier: Avantor
Description: Potassium dihydrogen phosphate 98.0-100.5% (dried basis) NF, FCC, J.T.Baker®
Catalog Number: (VWRC153184U)
Supplier: VWR Chemicals
Description: Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC
UOM: 1 * 500 g

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR at sales@sg.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
1 - 16 of 36
no targeter for Bottom