Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol : Chloroform : iso-Amyl alcohol (25:24:1) in saturated aqueous solution, pH 6.7, with alkaline buffer

Supplier: Alfa Aesar

Phenol:Chloroform:Isoamyl alcohol 25:24:1 is used in the extraction of DNA for the generation of DNA minisatellite fingerprints. It is also used for purifying RNA.

pH 6,7 with alkaline buffer.


CAS Number: 136112-00-0
Specification Test Results
Form Liquid
Solubility Soluble in water

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
100 ml ALFAJ60331.AE
Supplier Article Number:: J60331.AE
400 ml ALFAJ60331.AN
Supplier Article Number:: J60331.AN
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty