Test kit, AF 353, Lovibond®
Supplier: Tintometer
TINT413530EA 1270 SGD
TINT413530
Test kit, AF 353, Lovibond®
The AF 353 test kit is designed for estimation of blood haemoglobin levels using Harrison's method.
Blood is run directly into a special capillary blood cell and the colour of the blood is matched in a comparator instrument with a series of precalibrated glass filters. The filters are housed in a pair of test discs: Disc 5/8A covers 20 – 58 % haemoglobin; disc 5/8B covers 64 – 130 % haemoglobin (100 % is equivalent to 14,6 g per 100 ml).