Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

UV/Vis biophotometers, Genesys™ 10S

Supplier: Thermo Scientific
The Genesys 10S are UV/visible biophotometers with improved performance.

  • Bandwidth for improved nucleic acid measurement (ratio and concentration)
  • Direct protein measurement at 280 and 205 nm
  • Standard measurement of protein concentrations using Coomassie/Bradford; Lowry; modified Lowry; BCA; Pierce Micro-BCA and 660 Protein Assay
  • Six cell changer for automated multi-sample experiments

Versatile and intuitive with powerful internal software, preconfigured methods that can be edited and stored to a customised assay.

Delivery information: Supplied with a six position cell changer, single cell holder, spare fuses, USB memory device, USB cable, dust cover and operator manual.

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Description Range Pk VWR Catalog Number Quantity
UV/Vis biophotometer with power leads, EU- and UK-plug 190 − 1100 nm 1 SPET840-209300
Supplier Article Number:: 840-209300
UV-Vis biophotometer with power leads and internal printer, EU- and UK-plug - 1 SPET840-209400
Supplier Article Number:: 840-209400


Light source Pulsed Xenon
Spectral bandwidth 1,8 nm
Absorbance range 0,000 − 3,500 A
Absorbance accuracy ±0,005 A @ 260 nm
Photometric accuracy 0,08 % T
Stray light 0,08 % T
Wavelength range 190 − 1100 nm
Scan speed 10 - 4200 nm per minute
Display Backlit graphical LCD (91×71 mm) − 320×240 p
Electrical 100 - 240 V 100 − 240 V
W×D×H 300×400×250 nm
Weight 8,6 kg


Expand All / Collapse All

Description Pk VWR Catalog Number Quantity
Nanocell, 1.0 mm path length, 0,7 µl 1 SPET222-227800 Blocked
Supplier Article Number:: 222-227800
Paper for internal printer 5 Roll SPET335954-000 Blocked
Supplier Article Number:: 335954-000
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty