Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molecular sieve A4 (0.4 nm, 4 Å), extruders TECHNICAL, Ø 1.6

Supplier: VWR Chemicals

Adsorption of molecules smaller than 4 µm in diameter


Density: 1,1-2,2g/cm³ (20 °C)
MDL Number: MFCD00147627
CAS Number: 70955-01-0
EINECS: 215-283-8
UN: 0000
Specification Test Results
Identification Passes test

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Bucket (Plastic) 5 kg VWRC28464.364
Plastic bottle for solids 1 kg VWRC28464.295
Get a discount if you order 6 or more. Login in now for details.
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty